3-Bromo-5-(1-hydroxyethyl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

23645-1G 23645-25G 23645-5G
76885-274EA 84.88 USD
76885-274 76885-276 76885-278
3-Bromo-5-(1-hydroxyethyl)pyridine ≥97% research grade
3-Bromo-5-(1-hydroxyethyl)pyridine
Formula: C₇H₈BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD13188109
CAS Number: 283608-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR