1-Iodo-3-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

23627-1G 23627-25G 23627-5G
76885-264EA 79.57 USD
76885-264 76885-266 76885-268
1-Iodo-3-methoxy-2-methylbenzene ≥97%
1-Iodo-3-methoxy-2-methylbenzene
Formula: C₈H₉IO
MW: 248.0609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01833807
CAS Number: 21093-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR