(6-(Dimethylamino)-4-methylpyridin-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Dimethylamino)-4-methylpyridine-5-boronic acid

23500-1G 23500-25G 23500-500MG 23500-5G
76885-236EA 213.25 USD
76885-236 76885-238 76885-240 76885-242
(6-(Dimethylamino)-4-methylpyridin-3-yl)boronic acid ≥97%
(6-(Dimethylamino)-4-methylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₃BN₂O₂
MW: 180.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12912694
CAS Number: 535934-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR