2,3-Difluoro-5-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23406-1G 23406-25G 23406-500MG 23406-5G
76885-212EA 144.29 USD
76885-212 76885-214 76885-216 76885-218
2,3-Difluoro-5-hydroxybenzoic acid ≥97%
2,3-Difluoro-5-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08704572
CAS Number: 749230-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR