2,3-Difluoro-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23342-25G 23342-5G
76885-202EA 346.92 USD
76885-202 76885-204
2,3-Difluoro-6-methoxybenzoic acid ≥97%
2,3-Difluoro-6-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD04116018
CAS Number: 773873-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR