(3-Bromo-6-methoxy-2-methylphenyl)amine ≥95%

Supplier: AOBChem USA

Warning

23262-1G 23262-25G 23262-500MG 23262-5G
76885-174EA 367.08 USD
76885-174 76885-176 76885-178 76885-180
(3-Bromo-6-methoxy-2-methylphenyl)amine ≥95%
(3-Bromo-6-methoxy-2-methylphenyl)amine
Formula: C₈H₁₀BrNO
Storage Temperature: Ambient
CAS Number: 786596-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR