1-(4-Bromobenzyl)piperidine ≥97% research grade

Supplier: AOBChem USA

Warning

23216-25G 23216-5G
76885-166EA 310.85 USD
76885-166 76885-168
1-(4-Bromobenzyl)piperidine ≥97% research grade
1-(4-Bromobenzyl)piperidine
Formula: C₁₂H₁₆BrN
Storage Temperature: Ambient
MDL Number: MFCD06080709
CAS Number: 178162-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR