1,3-Difluoro-2-isopropoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 1,3-Difluoro-2-(1-methylethoxy)benzene

23022-1G 23022-25G 23022-5G
76885-104EA 62.6 USD
76885-104 76885-106 76885-108
1,3-Difluoro-2-isopropoxybenzene ≥97% research grade
1,3-Difluoro-2-isopropoxybenzene
Formula: C₉H₁₀F₂O
MW: 172.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056750
CAS Number: 1219020-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR