1-Bromo-2-fluoro-4,5-dimethoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

22998-25G 22998-5G
76885-094EA 488.02 USD
76885-094 76885-096
1-Bromo-2-fluoro-4,5-dimethoxybenzene ≥97% research grade
1-Bromo-2-fluoro-4,5-dimethoxybenzene
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17237566
CAS Number: 1095544-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR