(3'-methyl-[1,1'-biphenyl]-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

22907-1G 22907-25G 22907-5G
76885-080EA 114.58 USD
76885-080 76885-082 76885-084
(3'-methyl-[1,1'-biphenyl]-4-yl)boronic acid ≥97%
(3'-methyl-[1,1'-biphenyl]-4-yl)boronic acid
Formula: C₁₃H₁₃BO₂
Storage Temperature: Ambient
MDL Number: MFCD08701725
CAS Number: 501944-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR