(6-Propoxynaphthalen-2-yl)boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
22500-25G
22500-5G
76885-044EA
488.02
USD
76885-044
76885-046
(6-Propoxynaphthalen-2-yl)boronic acid ≥97%
(6-Propoxynaphthalen-2-yl)boronic acid
Formula:
C₁₃H₁₅BO₃ MW: 230.067 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16260718 CAS Number: 1228309-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...