2,3,4-Trifluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22482-1G 22482-25G 22482-500MG 22482-5G
76885-028EA 168.69 USD
76885-028 76885-030 76885-032 76885-034
2,3,4-Trifluoro-5-methylbenzoic acid ≥97%
2,3,4-Trifluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD20643360
CAS Number: 65829-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR