1-Methoxy-3-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

22325-25G 22325-5G
76885-000EA 502.87 USD
76885-000 76885-002
1-Methoxy-3-(methoxymethoxy)benzene ≥97%
1-Methoxy-3-(methoxymethoxy)benzene
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30473851
CAS Number: 57234-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR