1-Bromo-4-(2-bromoethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-(2-Bromoethexy)benzene, 1-(2-Bromoethoxy-4-bromobenzene)

22212-25G 22212-5G
76884-970EA 307.67 USD
76884-970 76884-972
1-Bromo-4-(2-bromoethoxy)benzene ≥97%
1-Bromo-4-(2-bromoethoxy)benzene
Formula: C₈H₈Br₂O
MW: 279.959 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044201
CAS Number: 18800-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR