2,4-Dibromo-6-fluorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

22115-1G 22115-25G 22115-500MG 22115-5G
76884-958EA 312.97 USD
76884-958 76884-960 76884-962 76884-964
2,4-Dibromo-6-fluorobenzyl alcohol ≥95%
2,4-Dibromo-6-fluorobenzyl alcohol
Formula: C₇H₅Br₂FO
MW: 283.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094090
CAS Number: 497181-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR