(5-Bromo-2-fluoropyridin-3-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

22092-1G 22092-25G 22092-500MG 22092-5G
76884-942EA 165.51 USD
76884-942 76884-944 76884-946 76884-948
(5-Bromo-2-fluoropyridin-3-yl)methanol ≥95%
(5-Bromo-2-fluoropyridin-3-yl)methanol
Formula: C₆H₅BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD15528904
CAS Number: 1227601-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR