(2,3-Dichloro-4-pyridinyl)methanol ≥97%

Supplier: AOBChem USA

Warning

22088-1G 22088-25G 22088-500MG 22088-5G
76884-926EA 203.7 USD
76884-926 76884-928 76884-930 76884-932
(2,3-Dichloro-4-pyridinyl)methanol ≥97%
(2,3-Dichloro-4-pyridinyl)methanol
Formula: C₆H₅Cl₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD14581635
CAS Number: 329794-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR