1-Bromo-3-isopropoxy-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

21956-1G 21956-25G 21956-500MG 21956-5G
76884-896EA 173.99 USD
76884-896 76884-898 76884-900 76884-902
1-Bromo-3-isopropoxy-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-isopropoxy-5-(trifluoromethyl)benzene
Formula: C₁₀H₁₀BrF₃O
MW: 283.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356474
CAS Number: 1369784-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR