1-Butylpyrazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Butyl-1H-pyrazole

21912-25G 21912-5G
76884-878EA 410.57 USD
76884-878 76884-880
1-Butylpyrazole ≥97%
1-Butylpyrazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MW: 124.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08701137
CAS Number: 52096-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR