2-(Benzyloxy)-3-fluorophenol ≥95%

Supplier: AOBChem USA

Warning

21799-1G 21799-25G 21799-500MG 21799-5G
76884-854EA 189.91 USD
76884-854 76884-856 76884-858 76884-860
2-(Benzyloxy)-3-fluorophenol ≥95%
2-(Benzyloxy)-3-fluorophenol
Formula: C₁₃H₁₁FO₂
MW: 218.224 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24715292
CAS Number: 148872-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR