1-Bromo-3-fluoro-2-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

21605-1G 21605-25G 21605-500MG 21605-5G
76884-806EA 189.91 USD
76884-806 76884-808 76884-810 76884-812
1-Bromo-3-fluoro-2-methoxy-4-methylbenzene ≥97%
1-Bromo-3-fluoro-2-methoxy-4-methylbenzene
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD19105383
CAS Number: 1239591-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR