2-(3-Bromo-5-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

21104-1G 21104-25G 21104-500MG 21104-5G
76884-706EA 225.98 USD
76884-706 76884-708 76884-710 76884-712
2-(3-Bromo-5-methoxyphenyl)-1,3-dioxolane ≥97%
2-(3-Bromo-5-methoxyphenyl)-1,3-dioxolane
Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619411
CAS Number: 2221811-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR