(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: N-Boc-1H-pyrazole-4-boronic acid, 1-tert-Butoxycarbonyl-1H-pyrazole-4-boronic acid, 1H-Pyrazole-4-boronic acid, N1-BOC protected

21077-25G 21077-5G
76884-690EA 381.93 USD
76884-690 76884-692
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid ≥95% research grade
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid
Formula: C₈H₁₃BN₂O₄
Storage Temperature: Ambient
MDL Number: MFCD09951915
CAS Number: 1188405-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR