(4-Chloro-3-ethoxy-2-fluorophenyl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

21003-25G 21003-5G
76884-680EA 381.93 USD
76884-680 76884-682
(4-Chloro-3-ethoxy-2-fluorophenyl)boronic acid ≥95% research grade
(4-Chloro-3-ethoxy-2-fluorophenyl)boronic acid
Formula: C₈H₉BClFO₃
MW: 218.418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015737
CAS Number: 1256346-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR