(2,6-Difluoro-3-pyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2,6-Difluoro-3-pyridyl)methanol

20886-1G 20886-25G 20886-500MG 20886-5G
76884-668EA 119.89 USD
76884-668 76884-670 76884-672 76884-674
(2,6-Difluoro-3-pyridin-3-yl)methanol ≥97%
(2,6-Difluoro-3-pyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₅F₂NO
MW: 145.107 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12911481
CAS Number: 1105510-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR