2-(1-Pyrrolidinylmethyl)phenylbronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

20752-1G 20752-25G 20752-500MG 20752-5G
76884-638EA 177.18 USD
76884-638 76884-640 76884-642 76884-644
2-(1-Pyrrolidinylmethyl)phenylbronic acid ≥97% research grade
2-(1-Pyrrolidinylmethyl)phenylbronic acid
Formula: C₁₁H₁₆BNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06801714
CAS Number: 878289-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR