1,3-Dibromo-2-(1-methylethoxy)-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

20692-1G 20692-25G 20692-5G
76884-624EA 62.6 USD
76884-624 76884-626 76884-628
1,3-Dibromo-2-(1-methylethoxy)-5-methylbenzene ≥97%
1,3-Dibromo-2-(1-methylethoxy)-5-methylbenzene
Formula: C₁₀H₁₂Br₂O
Storage Temperature: Refrigerator
MDL Number: MFCD19105369
CAS Number: 1310416-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR