2,3-Dibromoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,2-Dibromo-3-methoxybenzene, 2,3-Dibromophenyl methyl ether

20635-1G 20635-25G 20635-5G
76884-610EA 94.43 USD
76884-610 76884-612 76884-614
2,3-Dibromoanisole ≥97%
2,3-Dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864854
CAS Number: 95970-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR