2,4-Dichloro-3-methoxybenzoic acid 95% research grade

Supplier: AOBChem USA

Warning

20442-1G 20442-500MG 20442-5G 20442-25G
76884-588EA 471.04 USD
76884-588 76884-590 76884-592 77657-938
2,4-Dichloro-3-methoxybenzoic acid 95% research grade
2,4-Dichloro-3-methoxybenzoic acid
Formula: C₈H₆Cl₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD26960583
CAS Number: 115382-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR