1-Benzyloxy-4-bromo-2,5-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

20425-25G 20425-5G
76884-580EA 346.92 USD
76884-580 76884-582
1-Benzyloxy-4-bromo-2,5-difluorobenzene ≥97%
1-Benzyloxy-4-bromo-2,5-difluorobenzene
Formula: C₁₃H₉BrF₂O
MW: 299.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26385141
CAS Number: 901238-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR