1-Bromo-2-fluoro-4-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

20406-25G 20406-5G
76884-570EA 531.52 USD
76884-570 76884-572
1-Bromo-2-fluoro-4-(methoxymethoxy)benzene ≥97%
1-Bromo-2-fluoro-4-(methoxymethoxy)benzene
Formula: C₈H₈BrFO₂
MW: 235.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21195354
CAS Number: 1054314-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR