1,3,5-Tribromo-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

20377-1G 20377-25G 20377-5G
76884-560EA 84.88 USD
76884-560 76884-562 76884-564
1,3,5-Tribromo-2-iodobenzene ≥97%
1,3,5-Tribromo-2-iodobenzene
Formula: C₆H₂Br₃I
MW: 440.697 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00019010
CAS Number: 21521-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR