1-Bromo-3-butoxy-5-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

20346-1G 20346-25G 20346-500MG 20346-5G
76884-548EA 162.32 USD
76884-548 76884-550 76884-552 76884-554
1-Bromo-3-butoxy-5-(trifluoromethoxy)benzene ≥97%
1-Bromo-3-butoxy-5-(trifluoromethoxy)benzene
Formula: C₁₁H₁₂BrF₃O₂
MW: 313.111 g/mol
Storage Temperature: Ambient
CAS Number: 1221658-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR