1-(Benzyloxy)-3-bromo-5-(trifluoromethoxy)benzene 97%

Supplier: AOBChem USA

Warning

20343-1G 20343-25G 20343-5G 20343-500MG
76884-534EA 100.79 USD
76884-534 76884-536 76884-538 77649-218
1-(Benzyloxy)-3-bromo-5-(trifluoromethoxy)benzene 97%
1-(Benzyloxy)-3-bromo-5-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O₂
MW: 347.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036126
CAS Number: 1221658-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR