1-Bromo-4-(Methylsulfanyl)-2-(Trifluoromethyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

20304-1G 20304-25G 20304-500MG 20304-5G
76884-522EA 142.17 USD
76884-522 76884-524 76884-526 76884-528
1-Bromo-4-(Methylsulfanyl)-2-(Trifluoromethyl)Benzene ≥97%
1-Bromo-4-(Methylsulfanyl)-2-(Trifluoromethyl)Benzene
Formula: C₈H₆BrF₃S
MW: 271.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15527240
CAS Number: 1379356-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR