1,4-Diiodo-2-nitrobenzene ≥95%

Supplier: AOBChem USA

Warning

20287-1G 20287-25G 20287-5G
76884-508EA 62.6 USD
76884-508 76884-510 76884-512
1,4-Diiodo-2-nitrobenzene ≥95%
1,4-Diiodo-2-nitrobenzene
Formula: C₆H₃I₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD09030977
CAS Number: 89488-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR