2,3-Difluoro-4-hydroxybenzeneboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
20277-1G
20277-25G
20277-500MG
20277-5G
76884-492EA
319.34
USD
76884-492
76884-494
76884-496
76884-498
2,3-Difluoro-4-hydroxybenzeneboronic acid pinacol ester ≥97%
2,3-Difluoro-4-hydroxybenzeneboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BF₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD22414554 CAS Number: 1029439-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...