1-Bromo-3-(methoxymethyl)benzene ≥95% research grade

Supplier: AOBChem USA

Warning

20182-25G 20182-5G
76884-472EA 453.01 USD
76884-472 76884-474
1-Bromo-3-(methoxymethyl)benzene ≥95% research grade
1-Bromo-3-(methoxymethyl)benzene
Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD11504955
CAS Number: 1515-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR