2-(3-Chloro-5-fluorophenyl)-1,3-dioxolane 95%

Supplier: AOBChem USA

Warning

20164-1G 20164-25G 20164-5G 20164-500MG
76884-460EA 87 USD
76884-460 76884-462 76884-464 77645-140
2-(3-Chloro-5-fluorophenyl)-1,3-dioxolane 95%
2-(3-Chloro-5-fluorophenyl)-1,3-dioxolane
Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209957
CAS Number: 773086-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR