1-Bromo-4-chloro-2-fluoro-3-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

20162-1G 20162-25G 20162-500MG 20162-5G
76884-444EA 332.07 USD
76884-444 76884-446 76884-448 76884-450
1-Bromo-4-chloro-2-fluoro-3-ethoxybenzene ≥97%
1-Bromo-4-chloro-2-fluoro-3-ethoxybenzene
Formula: C₈H₇BrClFO
MW: 253.496 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28402072
CAS Number: 943830-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR