6-Bromo-2,3-difluorotoluene ≥97%

Supplier: AOBChem USA

Warning

19975-25G 19975-5G
76884-396EA 381.93 USD
76884-396 76884-398
6-Bromo-2,3-difluorotoluene ≥97%
6-Bromo-2,3-difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
Storage Temperature: Ambient
MDL Number: MFCD07777161
CAS Number: 847502-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR