1-Bromo-3-chloro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

19931-25G 19931-5G
76884-372EA 212.18 USD
76884-372 76884-374
1-Bromo-3-chloro-2-iodobenzene ≥97%
1-Bromo-3-chloro-2-iodobenzene
Formula: C₆H₃BrClI
MW: 317.348 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907935
CAS Number: 450412-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR