1-Bromo-3-chloro-5-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

19908-1G 19908-25G 19908-500MG 19908-5G
76884-358EA 155.96 USD
76884-358 76884-360 76884-362 76884-364
1-Bromo-3-chloro-5-(methoxymethoxy)benzene ≥97%
1-Bromo-3-chloro-5-(methoxymethoxy)benzene
Formula: C₈H₈BrClO₂
MW: 251.505 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619405
CAS Number: 2023712-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR