1-Bromo-4-butoxy-3,5,-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

19901-1G 19901-25G 19901-5G
76884-346EA 88.06 USD
76884-346 76884-348 76884-350
1-Bromo-4-butoxy-3,5,-dichlorobenzene ≥97%
1-Bromo-4-butoxy-3,5,-dichlorobenzene
Formula: C₁₀H₁₁BrCl₂O
MW: 298.004 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17011984
CAS Number: 1242070-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR