3-Bromo-2-chloroanisole ≥95%

Supplier: AOBChem USA

Warning

19598-25G 19598-5G
76884-318EA 233.4 USD
76884-318 76884-320
3-Bromo-2-chloroanisole ≥95%
3-Bromo-2-chloroanisole
Formula: C₇H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD16036127
CAS Number: 174913-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR