2-Benzyloxy-5-bromo-1,3-dimethylbenzene ≥97% research grade

Supplier: AOBChem USA

Warning

19521-25G 19521-5G
76884-312EA 346.92 USD
76884-312 76884-314
2-Benzyloxy-5-bromo-1,3-dimethylbenzene ≥97% research grade
2-Benzyloxy-5-bromo-1,3-dimethylbenzene
Formula: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD12913972
CAS Number: 168196-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR