1-Bromo-2-methyl-3-(phenylmethoxy)-benzene ≥97%

Supplier: AOBChem USA

Warning

19473-1G 19473-25G 19473-500MG 19473-5G
76884-294EA 126.25 USD
76884-294 76884-296 76884-298 76884-300
1-Bromo-2-methyl-3-(phenylmethoxy)-benzene ≥97%
1-Bromo-2-methyl-3-(phenylmethoxy)-benzene
Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30296477
CAS Number: 909032-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR