1-Bromo-2,3,4-trimethoxybenzene ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
19413-1G
19413-25G
19413-5G
76884-268EA
62.6
USD
76884-268
76884-270
76884-272
1-Bromo-2,3,4-trimethoxybenzene ≥97% research grade
1-Bromo-2,3,4-trimethoxybenzene
Formula:
C₉H₁₁BrO₃ MW: 247.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01318955 CAS Number: 10385-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...