1-Bromo-4-methoxy-2-phenylmethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

19362-1G 19362-25G 19362-500MG 19362-5G
76884-254EA 138.98 USD
76884-254 76884-256 76884-258 76884-260
1-Bromo-4-methoxy-2-phenylmethoxybenzene ≥95%
1-Bromo-4-methoxy-2-phenylmethoxybenzene
Formula: C₁₄H₁₃BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07367815
CAS Number: 150356-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR