1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-(Tert-butoxycarbonyl)-5-(4,4,5,5-Tetramethyl-[1,3,2]Dioxaborolan-2-yl)indazole, 1-Boc-1H-indalzole-5-boronic acid pinacol ester, tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
19297-1G
19297-25G
19297-5G
76884-244EA
62.6
USD
76884-244
76884-246
76884-248
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected ≥97%
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
Precaución: for Research Use Only
Formula:
C₁₈H₂₅BN₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD08437629 CAS Number: 864771-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...